missing translation for 'onlineSavingsMsg'
Learn More

10,16-Dihydroxyhexadecanoic Acid, TRC

Product Code. 30382830
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382830 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382830 Supplier Toronto Research Chemicals Supplier No. TRCD45393050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10,16-Dihydroxyhexadecanoic Acid
CAS 3233-90-7
Purity Grade Notes HPLC
Molecular Formula C16 H32 O4
Quantity 50 mg
InChI Formula InChI=1S/C16H32O4/c17-14-10-6-5-8-12-15(18)11-7-3-1-2-4-9-13-16(19)20/h15,17-18H,1-14H2,(H,19,20)
SMILES OCCCCCCC(O)CCCCCCCCC(=O)O
IUPAC Name 10,16-dihydroxyhexadecanoic acid
Molecular Weight (g/mol) 288.423
Formula Weight 288.23
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.