missing translation for 'onlineSavingsMsg'
Learn More

10,11-Epoxide Carbamazepine- d10, TRC

Product Code. 30366842
Change view
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366842 0.5 mg 0.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30366842 Supplier Toronto Research Chemicals Supplier No. TRCC1758560.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Carbamazepine 10,11-epoxide ring-d10
CAS 1219804-16-6
Molecular Formula C15 D10 H2 N2 O2
Quantity 0.5 mg
Synonym Carbamazepine-10,11-epoxide-d10, Carbamazepine 10,11-epoxide D10: 1a,10b-Dihydro-6H-dibenz[b,f]oxireno[d]azepine-6-carboxamide-1a,2,3,4,5,7,8,9,10.10b-d10, 10,11-Dihydro-10,11-epoxycarbamazepine-d10, 10,11-Dihydro-5H-dibenz[b,f]azepine-5-carboxamide-d10 10,11-epoxide
InChI Formula InChI=1S/C15H12N2O2/c16-15(18)17-11-7-3-1-5-9(11)13-14(19-13)10-6-2-4-8-12(10)17/h1-8,13-14H,(H2,16,18)/i1D,2D,3D,4D,5D,6D,7D,8D,13D,14D
SMILES [2H]c1c([2H])c([2H])c2c(N(C(=O)N)c3c([2H])c([2H])c([2H])c([2H])c3C4([2H])OC24[2H])c1[2H]
IUPAC Name 2,4,6,7,8,9,13,14,15,16-decadeuterio-3-oxa-11-azatetracyclo[10.4.0.0^{2,4}.0^{5,10}]hexadeca-1(12),5(10),6,8,13,15-hexaene-11-carboxamide
Molecular Weight (g/mol) 262.3296
Formula Weight 262.1526
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.