missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dihydro-5H-dibenzo[b,f]azepine (Iminodibenzyl), Mikromol™

Product Code. 17971456
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17971456 100 mg 100mg
17952846 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17971456 Supplier Mikromol Supplier No. MM0076.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

10,11-Dihydro-5H-dibenzo[b,f]azepine (Iminodibenzyl)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Iminodibenzyl (10,11-Dihydro-5H-dibenzo[b,f]azepine)
CAS 494-19-9
Analyte or Component Names Iminodibenzyl (10,11-Dihydro-5H-dibenzo[b,f]azepine)
Shipping Condition Room Temperature
Molecular Formula C14 H13 N
Certifications/Compliance ISO 17025
Synonym Trimipramine Maleate Imp. B (EP), Trimipramine Imp. B (EP), Iminodibenzyl, 10,11-Dihydro-5H-dibenzo[b,f]azepine, Clomipramine Hydrochloride Imp. E (EP), Trimipramine Maleate Imp. F (EP), Carbamazepine Imp. E (EP), Carbamazepine Impurity E, Clomipramine Hydrochloride Impurity E, Trimipramine Maleate Impurity F, Clomipramine Impurity E, Trimipramine Impurity F
InChI Formula InChI=1S/C14H13N/c1-3-7-13-11(5-1)9-10-12-6-2-4-8-14(12)15-13/h1-8,15H,9-10H2
SMILES C1Cc2ccccc2Nc3ccccc13
IUPAC Name 6,11-dihydro-5H-benzo[b][1]benzazepine
Molecular Weight (g/mol) 195.26
Quantity 100 mg
Formula Weight 195.1048 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.