missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dihydro-5H-dibenzo[b,f]-azepine-5-carboxamide (10,11-Dihydro-carbamazepine), Mikromol™

Product Code. 17984226
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17984226 25 mg 25mg
17903556 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17984226 Supplier Mikromol Supplier No. MM0076.040025
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

10,11-Dihydro-5H-dibenzo[b,f]-azepine-5-carboxamide (10,11-Dihydro-carbamazepine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10,11-Dihydro-carbamazepine
CAS 3564-73-6
Analyte or Component Names 10,11-Dihydro-carbamazepine
Shipping Condition Room Temperature
Molecular Formula C15 H14 N2 O
Certifications/Compliance ISO 17025
Synonym Carbamazepine Imp. A (EP), Carbamazepine USP Related Compound A, Carbamazepine USP RC A, 10,11-Dihydro-5H-dibenzo[b,f]-azepine-5-carboxamide, 10,11-Dihydro-carbamazepine, Carbamazepine Related Compound A, Carbamazepine Impurity A
InChI Formula InChI=1S/C15H14N2O/c16-15(18)17-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)17/h1-8H,9-10H2,(H2,16,18)
SMILES NC(=O)N1c2ccccc2CCc3ccccc13
IUPAC Name 5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
Molecular Weight (g/mol) 238.28
Quantity 25 mg
Formula Weight 238.1106 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.