missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-one (Dibenzosuberone), Mikromol™

Product Code. 17932716
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17932716 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17932716 Supplier Mikromol Supplier No. MM0062.01
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-one (Dibenzosuberone)
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Dibenzosuberone
CAS 1210-35-1
Analyte or Component Names Dibenzosuberone
Shipping Condition Room Temperature
Molecular Formula C15 H12 O
Certifications/Compliance ISO 17025
Synonym Amitriptyline Hydrochloride Imp. A (EP), Amitriptyline Imp. A (EP), Cyproheptadine Hydrochloride Imp. B (EP), Cyproheptadine Imp. B (EP), Nortriptyline Hydrochloride Imp. A (EP), Nortriptyline Imp. A (EP), Amitriptyline USP Related Compound A, Amitriptyline USP RC A, 10,11-Dihydro-5H-dibenzo[a,d][7]annulen-5-one, Dibenzosuberone, Amitriptyline Related Compound A, Amitriptyline Hydrochloride Impurity A, Cyproheptadine Hydrochloride Impurity B, Nortriptyline Hydrochloride Impurity A, Amitriptyline Impurity A, Cyproheptadine Impurity B, Nortriptyline Impurity A
InChI Formula InChI=1S/C15H12O/c16-15-13-7-3-1-5-11(13)9-10-12-6-2-4-8-14(12)15/h1-8H,9-10H2
SMILES O=C1c2ccccc2CCc3ccccc13
IUPAC Name 5,6-dihydrodibenzo[2,1-b:1',2'-e][7]annulen-11-one
Molecular Weight (g/mol) 208.26
Quantity 100 mg
Formula Weight 208.0888 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.