missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dihydro-10-hydroxy Carbamazepine, TRC

Product Code. 30388675
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30388675 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30388675 Supplier Toronto Research Chemicals Supplier No. TRCD44913510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10,11-Dihydro-10-Hydroxycarbamazepine
CAS 29331-92-8
Purity Grade Notes HPLC
Molecular Formula C15 H14 N2 O2
Quantity 10 mg
Synonym 10-Hydroxy-10,11-dihydrocarbamazepine, 10,11-Dihydro-10-hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, 10,11-Dihydro-10-hydroxycarbamazepine, 10-Hydroxy-10,11-dihydrocarbamezepine, BIA 2-005, GP 47779, Licarbazepine
InChI Formula InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)
SMILES NC(=O)N1c2ccccc2CC(O)c3ccccc13
IUPAC Name 5-hydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
Molecular Weight (g/mol) 254.28
Formula Weight 254.1055
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.