missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dehydrocurvularin, TRC

Product Code. 30357618
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
Unit Size:
2.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357618 5 mg 5mg
30359702 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30357618 Supplier Toronto Research Chemicals Supplier No. TRCD2303485MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 10,11-Dehydrocurvularin
CAS 1095588-70-7
Molecular Formula C20 H34 N2 O2
Quantity 5 mg
Synonym (4R,8E)-4,5,6,7-Tetrahydro-11,13-dihydroxy-4-methyl-2H-3-benzoxacyclododecin-2,10(1H)-dione, (+)-(10E,15R)-10,11-Dehydrocurvularin, ent-Dehydrocurvularin
InChI Formula InChI=1S/C20H34N2O2/c1-4-5-6-7-8-9-10-11-12-22(3)20-14-18(16-23)13-19(15-20)21-17(2)24/h13-15,23H,4-12,16H2,1-3H3,(H,21,24)
SMILES CC1CCCC=CC(=O)C2=C(CC(=O)O1)C=C(C=C2O)O
IUPAC Name N-[3-[decyl(methyl)amino]-5-(hydroxymethyl)phenyl]acetamide
Molecular Weight (g/mol) 334.496
Formula Weight 334.262
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.