missing translation for 'onlineSavingsMsg'
Learn More

10,11-Dehydro Misoprostol(Mixture of Diastereomers), TRC

Product Code. 30365707
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
50 mg
Unit Size:
2.5mg
25mg
50mg
Product Code. Quantity unitSize
30365707 2.5 mg 2.5mg
30365391 25 mg 25mg
30356753 50 mg 50mg
3 options
This item is not returnable. View return policy

Product Code. 30365707

Brand: Toronto Research Chemicals TRCD2305902.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58682-86-3
Formula Weight 364.26
InChI Formula InChI=1S/C22H36O4/c1-4-5-16-22(2,25)17-10-11-18-14-15-20(23)19(18)12-8-6-7-9-13-21(24)26-3/h10-11,14-15,18-19,25H,4-9,12-13,16-17H2,1-3H3/b11-10+/t18-,19+,22?/m0/s1
IUPAC Name methyl 7-((1R,2S)-2-((E)-4-hydroxy-4-methyloct-1-en-1-yl)-5-oxocyclopent-3-en-1-yl)heptanoate
Molecular Formula C22H36O4
Molecular Weight (g/mol) 364.52
SMILES O=C(OC)CCCCCC[C@@H]1[C@@H](/C=C/CC(O)(C)CCCC)C=CC1=O
Synonym (13E)-(+/-)-16-Hydroxy-16-methyl-9-oxoprosta-10,13-dien-1-oic Acid Methyl Ester
Chemical Name or Material 10,11-Dehydro Misoprostol (mixture of diastereomers)

Specifications

Quantity 2.5 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.