missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Positive Displacement Pipette Tips

Product Code. 15719160 Shop All MP Biomedicals Products
Click to view available options
Quantity:
60 Pk.
Unit Size:
Pack of 60
This item is not returnable. View return policy

Product Code. 15719160

Brand: MP Biomedicals™ 07DP1035

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Racks of 60 pre-assembled, pre-sterilized Positive Displacement Pipette Tips. Compatible with Saftest platform.

  • Replacement tips for for 10 to 100 μL postive displacement pipettor, catalog number 07WS1075 pipettor
  • Capillaries and pistons are pre-assembled and presterilized
  • Come in racks of 60
TRUSTED_SUSTAINABILITY

Specifications

Capacity 10 to 100 μL
For Use With (Equipment) SafTest System
For Use With (Application) Lipid degradation and oxidation
Packaging Type Box
Product Type Positive displacement pipet tips
Quantity 60 Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.