missing translation for 'onlineSavingsMsg'
Learn More

1-Vinylnaphthalene, 95%, stab. with 4-tert-butylcatechol

Product Code. 15469125
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469125 1 g 1g
15479125 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469125 Supplier Thermo Scientific Alfa Aesar Supplier No. H51043.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 826-74-4 | C12H10 | 154.21 g/mol

1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Vinylnaphthalene is used as a precursor of polynuclear aromatic hydrocarbons in tobacco smoke. It is also used as pharmaceutical intermediate.

Solubility
Soluble in ethyl acetate, hexane and methanol.

Notes
Moisture sensitive. Incompatible with oxidizing agents, heat, light, ultraviolet radiation and free radical initiators.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 826-74-4
Molecular Formula C12H10
Molecular Weight (g/mol) 154.21
MDL Number MFCD00075766
InChI Key IGGDKDTUCAWDAN-UHFFFAOYSA-N
Synonym 1-vinylnaphthalene, vinylnaphthalene, 1-vinyl naphthalene, unii-pf20cj2k0n, pf20cj2k0n, 1-vinylphthalene, 1-vinyl-naphthalene, naphthalene, ethenyl, naphthalen-1-yl ethene, naphthalene, 1-ethenyl
PubChem CID 70004
ChEBI CHEBI:51327
IUPAC Name 1-ethenylnaphthalene
SMILES C=CC1=C2C=CC=CC2=CC=C1

Specifications

Density 1.04
Boiling Point 135°C to 138°C
Quantity 1 g
Sensitivity Moisture sensitive
Solubility Information Soluble in ethyl acetate,hexane and methanol.
Formula Weight 154.21
Percent Purity 95%
Chemical Name or Material 1-Vinylnaphthalene, stabilized with 4-tert-butylcatechol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.