missing translation for 'onlineSavingsMsg'
Learn More

1-Vinylimidazole (Stabilized with Hydroquinone), TRC

Product Code. 30454311
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30454311 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30454311 Supplier Toronto Research Chemicals Supplier No. TRCV75703510MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Vinylimidazole
CAS 1072-63-5
Molecular Formula C5 H6 N2
Quantity 10 mg
Synonym Imidazole','1-vinyl- (6CI,7CI,8CI), 1-Ethenyl-1H-imidazole, 1-Vinyl-1H-imidazole, 1-Vinylimidazole, N-Vinylimidazole, 1H-Imidazole','1-ethenyl-
InChI Formula InChI=1S/C5H6N2/c1-2-7-4-3-6-5-7/h2-5H,1H2
SMILES C=Cn1ccnc1
IUPAC Name 1-ethenylimidazole
Molecular Weight (g/mol) 94.114
Formula Weight 94.0531
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.