missing translation for 'onlineSavingsMsg'
Learn More

1-Undecyne, TRC

Product Code. 30456812
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
30456812 1 g 1g
30450616 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 30456812

Brand: Toronto Research Chemicals TRCU7889901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-Undecyne
CAS 2243-98-3
Molecular Formula C11H20
Quantity 1 g
Synonym NSC 87633
InChI Formula InChI=1S/C11H20/c1-3-5-7-9-11-10-8-6-4-2/h1H,4-11H2,2H3
SMILES CCCCCCCCCC#C
IUPAC Name undec-1-yne
Molecular Weight (g/mol) 152.28
Formula Weight 152.1565
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.