missing translation for 'onlineSavingsMsg'
Learn More

1-Undecene, 94.0%, For GC analysis, MP Biomedicals™

Product Code. 11449711 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 mL
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449711 5 mL 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11449711 Supplier MP Biomedicals Supplier No. 0215823805
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 821-95-4
Color Colorless
Assay Percent Range 0.94
Molecular Formula C11H22
Quantity 5 mL
Synonym 1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720
InChI Key DCTOHCCUXLBQMS-UHFFFAOYSA-N
SMILES CCCCCCCCCC=C
IUPAC Name undec-1-ene
Molecular Weight (g/mol) 154.297
PubChem CID 13190
ChEBI CHEBI:77444
Formula Weight 154.3
Physical Form Liquid
Chemical Name or Material 1-Undecene, For GC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.