missing translation for 'onlineSavingsMsg'
Learn More

1-Undecanol, 98%

Product Code. 10123670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10123670 500 mL Glass bottle 500mL
10686722 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10123670 Supplier Thermo Scientific Acros Supplier No. 140685000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-42-5 | C11H24O | 172.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-42-5
Molecular Formula C11H24O
Molecular Weight (g/mol) 172.31
MDL Number MFCD00004751
InChI Key KJIOQYGWTQBHNH-UHFFFAOYSA-N
Synonym 1-undecanol, undecyl alcohol, undecanol, n-undecanol, n-undecyl alcohol, hendecanoic alcohol, hendecyl alcohol, 1-hendecanol, n-hendecylenic alcohol, tip-nip
PubChem CID 8184
ChEBI CHEBI:87499
IUPAC Name undecan-1-ol
SMILES CCCCCCCCCCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 11.0°C
Color Colorless to Yellow
Density 0.8310g/mL
Boiling Point 146.0°C (30.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)10OH
Refractive Index 1.4370 to 1.4420
Quantity 500 mL
Beilstein 01, 427
Specific Gravity 0.831
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in 60% alcohol
Formula Weight 172.31
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Undecanol, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Undecanol
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.