missing translation for 'onlineSavingsMsg'
Learn More

1-Tritylimidazole, 98%, Thermo Scientific Chemicals

Product Code. 11368636
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11368636

Brand: Thermo Scientific Alfa Aesar L17694.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

It is a synthesis material intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a synthesis material intermediate.

Solubility
Insoluble in water.

Notes
Store in cool dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15469-97-3
Molecular Formula C22H18N2
Molecular Weight (g/mol) 310.40
MDL Number MFCD00229427
InChI Key NPZDCTUDQYGYQD-UHFFFAOYSA-N
Synonym 1-trityl-1h-imidazole, 1-triphenylmethyl imidazole, n-tritylimidazole, 1-triphenylmethyl-1h-imidazole, clotrimazole impurity f, triphenylmethyl imidazole, 1-triphenylmethylimidazole, 1-tritylimidazol, pubchem23733, acmc-209dbb
PubChem CID 618231
SMILES C1=CN(C=N1)C(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 220°C to 224°C
Odor Odorless
Quantity 1 g
Beilstein 238171
Solubility Information Insoluble in water.
IUPAC Name 1-tritylimidazole
Formula Weight 310.4
Percent Purity 98%
Chemical Name or Material 1-Tritylimidazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.