missing translation for 'onlineSavingsMsg'
Learn More

1-Trityl-L-histidine, 98%, Thermo Scientific Chemicals

Product Code. 15479258
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35146-32-8
Molecular Formula C25H23N3O2
Molecular Weight (g/mol) 397.478
MDL Number MFCD00153444
InChI Key BSZQZNOAYQCQFZ-QHCPKHFHSA-N
Synonym h-his trt-oh, 1-triphenylmethyl-l-histidine, l-histidine, 1-triphenylmethyl, s-2-amino-3-1-trityl-1h-imidazol-4-yl propanoic acid, 2s-2-amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, 1-trityl-l-histidine, pubchem18948, n-im-trityl-l-histidine, n im-trityl-l-histidine
PubChem CID 7408287
IUPAC Name (2S)-2-amino-3-(1-tritylimidazol-4-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)N

Specifications

Melting Point 210°C (decomposition)
Beilstein 767244
Formula Weight 397.47
Percent Purity 98%
Quantity 5 g
Physical Form Solid
Chemical Name or Material 1-Trityl-L-histidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'