missing translation for 'onlineSavingsMsg'
Learn More

1-Trimethylsilyl-2-pyrrolidinone, 96%, Thermo Scientific™

Product Code. 11377775
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377775 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11377775 Supplier Thermo Scientific Alfa Aesar Supplier No. L02638.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
1-Trimethylsilyl-2-pyrrolidinone, is used as a fine chemical intermediate.

Solubility
It hydrolyzes with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14468-90-7
Molecular Formula C7H15NOSi
Molecular Weight (g/mol) 157.29
MDL Number MFCD00051572
InChI Key LUBVCBITQHEVCJ-UHFFFAOYSA-N
Synonym 1-trimethylsilyl pyrrolidin-2-one, 1-trimethylsilyl-2-pyrrolidinone, n-trimethylsilyl-2-pyrrolidinone, n-trimethylsilyl-2-pyrrolidone, 1-trimethylsilanyl-pyrrolidin-2-one, 2-pyrrolidinone, 1-trimethylsilyl, acmc-20aorp, 1-trimethylsilylpyrrolidone, n-trimethylsilylpyrrolid-2-one
PubChem CID 84461
IUPAC Name 1-trimethylsilylpyrrolidin-2-one
SMILES C[Si](C)(C)N1CCCC1=O

Specifications

Density 0.821
Boiling Point 90°C (20 mmHg)
Flash Point 46°C (115°F)
Refractive Index 1.46
Quantity 50 g
UN Number UN1993
Beilstein 114950
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes with water.
Formula Weight 157.29
Percent Purity 96%
Chemical Name or Material 1-Trimethylsilyl-2-pyrrolidinone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.