missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsilyl)-1H-benzotriazole, 97%, Thermo Scientific™

Product Code. 11926641 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11926641 5 g 5g
11916641 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11926641 Supplier Thermo Scientific Acros Supplier No. 426260050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 43183-36-4
Molecular Formula C9H13N3Si
Molecular Weight (g/mol) 191.31
InChI Key VHOLOZYRMQVHBK-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1h-benzotriazole, 1-trimethylsilylbenzotriazole, 1-trimethylsilyl-1h-benzo d 1,2,3 triazole, 1-trimethylsilyl-1,2,3-benzotriazole, 1-tms-1h-benzotriazole, acmc-1bn8x, n-trimethylsilylbenzotriazole, benzotriazol-1-yl trimethyl silane, 1-trimethylsilanyl-1h-benzotriazole
PubChem CID 3558326
IUPAC Name benzotriazol-1-yl(trimethyl)silane
SMILES C[Si](C)(C)N1C2=CC=CC=C2N=N1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.0540g/mL
Boiling Point 125.0°C to 126.0°C (3.0 mmHg)
Flash Point −3°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.054
Formula Weight 191.31
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(Trimethylsilyl)-1H-benzotriazole
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-(Trimethylsilyl)-1H-benzotriazole
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.