missing translation for 'onlineSavingsMsg'
Learn More

1-Trimethylsilyl-1,4-pentadiyne, 98%

Product Code. 15430248
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15430248 25 g 25g
15410248 1 g 1g
15420248 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15430248 Supplier Thermo Scientific Alfa Aesar Supplier No. H53436.14
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 71789-10-1 | C8H12Si | 136.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Incompatible with strong oxidizing agents and ammonia.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71789-10-1
Molecular Formula C8H12Si
Molecular Weight (g/mol) 136.27
MDL Number MFCD00066350
InChI Key JKYMNPFNLMDSFG-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1,4-pentadiyne, trimethyl penta-1,4-diynyl silane, trimethyl penta-1,4-diyn-1-yl silane, 1,4-pentadiynyltrimethylsilane, silane,trimethyl-1,4-pentadiyn-1-yl
PubChem CID 144510
IUPAC Name trimethyl(penta-1,4-diynyl)silane
SMILES C[Si](C)(C)C#CCC#C

Specifications

Boiling Point 42°C (25mmHg)
Quantity 25 g
Flash Point 8°C (46°F)
Density 0.789
Linear Formula HC≡CCH2C≡CSi(CH3)3
UN Number UN1993
Formula Weight 136.27
Percent Purity 98%
Chemical Name or Material 1-Trimethylsilyl-1,4-pentadiyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.