missing translation for 'onlineSavingsMsg'
Learn More

1-(Trimethylsiloxy)cyclohexene, 98%

Product Code. 11443127
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443127 50 g 50g
11433127 10 g 10g
11453127 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11443127 Supplier Thermo Scientific Alfa Aesar Supplier No. A16099.18
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6651-36-1 | C9H18OSi | 170.33 g/mol

1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trimethylsiloxy)cyclohexene is used as intermediate to complex compounds, including natural products. It is also used as a pharmaceutical intermediate.

Solubility
Not miscible or difficult to mix.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6651-36-1
Molecular Formula C9H18OSi
Molecular Weight (g/mol) 170.33
MDL Number MFCD00001541
InChI Key SBEMOANGDSSPJY-UHFFFAOYSA-N
Synonym 1-trimethylsilyloxy cyclohexene, cyclohex-1-en-1-yloxy trimethylsilane, 1-trimethylsiloxy cyclohexene, silane, 1-cyclohexen-1-yloxy trimethyl, 1-cyclohexenyloxytrimethylsilane, 1-cyclohexen-1-yloxy trimethylsilane, cyclohexene, 1-trimethylsilyl oxy, cyclohexenyloxy trimethylsilane, 1-trimethylsiloxycyclohexene, cyclohexenyloxytrimethylsilane
PubChem CID 81161
IUPAC Name cyclohexen-1-yloxy(trimethyl)silane
SMILES C[Si](C)(C)OC1=CCCCC1

Specifications

Boiling Point 165°C
Quantity 50 g
Flash Point 42°C (107°F)
Refractive Index 1.447
Sensitivity Moisture sensitive
Density 0.883
UN Number UN1993
Beilstein 1859394
Solubility Information Not miscible or difficult to mix.
Formula Weight 170.33
Percent Purity 98%
Chemical Name or Material 1-(Trimethylsiloxy)cyclohexene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.