missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoromethyl)vinyl acetate, 97%

Product Code. 11469647
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469647 250 g 250g
11459647 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469647 Supplier Thermo Scientific Alfa Aesar Supplier No. B22133.30
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 2247-91-8 | C5H5F3O2 | 154.088 g/mol

1-(Trifluoromethyl)vinyl acetate is used as a chemical, organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trifluoromethyl)vinyl acetate is used as a chemical, organic Intermediates.

Solubility
Not miscible or difficult to mix in water.

Notes
Moisture sensitive. Store away from strong oxidizing agents, heat, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2247-91-8
Molecular Formula C5H5F3O2
Molecular Weight (g/mol) 154.088
MDL Number MFCD00040844
InChI Key VOKGSDIHTCTXDS-UHFFFAOYSA-N
Synonym 1-trifluoromethyl vinyl acetate, 1-trifluoromethyl ethenyl acetate, 1-propen-2-ol, 3,3,3-trifluoro-, acetate, 1-propen-2-ol, 3,3,3-trifluoro-, 2-acetate, trifluoroacetone enol acetate, trifluoroacetone enolacetate, alpha-trifluoromethylvinyl acetate, acmc-1cjli, 3,3,3-trifluoropropen-2-yl acetate, alpa-trifluoromethyl vinyl acetate
PubChem CID 75254
IUPAC Name 3,3,3-trifluoroprop-1-en-2-yl acetate
SMILES CC(=O)OC(=C)C(F)(F)F

Specifications

Density 1.2
Boiling Point 86°C to 88°C
Flash Point 2°C (35°F)
Odor Pungent
Refractive Index 1.342
Quantity 250 g
UN Number UN3272
Beilstein 2078331
Sensitivity Moisture sensitive
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 154.09
Percent Purity 97%
Chemical Name or Material 1-(Trifluoromethyl)vinyl acetate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.