missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoromethyl)cyclopentanecarboxylic acid, 97%

Product Code. 11341989
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341989 1 g 1g
15437545 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341989 Supplier Thermo Scientific Alfa Aesar Supplier No. H26055.03
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 277756-44-2 | C7H9F3O2 | 182.142 g/mol

1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-(Trifluoromethyl)cyclopentanecarboxylic acid is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 277756-44-2
Molecular Formula C7H9F3O2
Molecular Weight (g/mol) 182.142
MDL Number MFCD08445821
InChI Key DGQRYPPBAJNZFZ-UHFFFAOYSA-N
Synonym 1-trifluoromethyl cyclopentane-1-carboxylic acid, 1-trifluoromethyl cyclopentanecarboxylic acid, acmc-1cp3t, 1-trifluoromethyl-cyclopentanecarboxylic acid, 1-trifluoromethylcyclopentane carboxylic acid, cyclopentanecarboxylicacid, 1-trifluoromethyl
PubChem CID 15507525
IUPAC Name 1-(trifluoromethyl)cyclopentane-1-carboxylic acid
SMILES C1CCC(C1)(C(=O)O)C(F)(F)F

Specifications

Melting Point 36°C to 38°C
Quantity 1 g
Solubility Information Insoluble in water.
Formula Weight 182.15
Percent Purity 97%
Chemical Name or Material 1-(Trifluoromethyl)cyclopentanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.