missing translation for 'onlineSavingsMsg'
Learn More

1-(Trifluoroacetyl)imidazole, 98+%

Product Code. 11312675
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312675 50 g 50g
11302675 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312675 Supplier Thermo Scientific Alfa Aesar Supplier No. L00135.18
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1546-79-8 | C5H3F3N2O | 164.087 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1546-79-8
Molecular Formula C5H3F3N2O
Molecular Weight (g/mol) 164.087
MDL Number MFCD00014501
InChI Key SINBGNJPYWNUQI-UHFFFAOYSA-N
Synonym 1-trifluoroacetyl imidazole, n-trifluoroacetylimidazole, 1-trifluoroacetyl-1h-imidazole, 1h-imidazole, 1-trifluoroacetyl, unii-8c6041ow3i, tfai, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethanone, n-trifluoroacetyl imidazole, 2,2,2-trifluoro-1-imidazol-1-yl ethanone, 2,2,2-trifluoro-1-1h-imidazol-1-yl ethan-1-one
PubChem CID 73767
IUPAC Name 2,2,2-trifluoro-1-imidazol-1-ylethanone
SMILES C1=CN(C=N1)C(=O)C(F)(F)F

Specifications

Density 1.441
Boiling Point 45°C to 46°C (14 mmHg)
Flash Point 45°C (113°F)
Odor Pungent
Refractive Index 1.424
Quantity 50 g
UN Number UN1993
Beilstein 608904
Sensitivity Moisture sensitive; May form a precipitate
Formula Weight 164.09
Percent Purity ≥98%
Chemical Name or Material 1-(Trifluoroacetyl)imidazole
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.