missing translation for 'onlineSavingsMsg'
Learn More

1-Tridecyne, 97%

Product Code. 11337725
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11337725 5 g 5g
11327725 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11337725 Supplier Thermo Scientific Alfa Aesar Supplier No. L02481.06
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26186-02-7 | C13H24 | 180.34 g/mol

It finds its application in the synthesis of unsaturated fatty acids and in the stereoselective synthesis of d-erythro- and l-threo-sphinganines via palladium-catalyzed equilibration and Suzuki coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its application in the synthesis of unsaturated fatty acids and in the stereoselective synthesis of d-erythro- and l-threo-sphinganines via palladium-catalyzed equilibration and Suzuki coupling.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26186-02-7
Molecular Formula C13H24
Molecular Weight (g/mol) 180.34
MDL Number MFCD00015074
InChI Key GZEDKDBFUBPZNG-UHFFFAOYSA-N
Synonym 1-tridecyne, tridecyne, acmc-1cinm
PubChem CID 117754
IUPAC Name tridec-1-yne
SMILES CCCCCCCCCCCC#C

Specifications

Density 0.78
Boiling Point 116°C to 118°C (10 mmHg)
Flash Point 75°C (167°F)
Linear Formula CH3(CH2)10C≡CH
Refractive Index 1.438
Quantity 5 g
Beilstein 1744658
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 180.34
Percent Purity 97%
Chemical Name or Material 1-Tridecyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.