missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Thioglycerol, TRC
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Specifications
Specifications
| Chemical Name or Material | 1-Thioglycerol |
| CAS | 96-27-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C3 H8 O2 S |
| Quantity | 5 g |
| Synonym | 3-Mercapto-1,2-propanediol, (+/-)-3-Mercapto-1,2-propanediol, 1,2-Dihydroxy-3-mercaptopropane, 1-Mercapto-2,3-dihydroxypropane, 1-Mercapto-2,3-propanediol, 1-Mercaptoglycerol, 1-Monothioglycerol, 1-Thio-2,3-propanediol, 1-Thio-DL-glycerol, 1-Thioglycerol, 2,3-Dihydroxy-1-propanethiol, 2,3-Dihydroxypropanethiol, 3-Mercapto-1,2-dihydroxypropane, DL-1-Thioglycerol, Glycerol-1-thiol, NSC 5370, Thioglycerin, Thioglycerol, Thiovanol, alpha-Monothioglycerol, alpha-Thioglycerin, alpha-Thioglycerol, alpha-Thiolglycerol, (±)-3-Mercapto-1,2-propanediol, α-Monothioglycerol, α-Thioglycerin, α-Thioglycerol, α-Thiolglycero |
| InChI Formula | InChI=1S/C3H8O2S/c4-1-3(5)2-6/h3-6H,1-2H2 |
| SMILES | OCC(O)CS |
| IUPAC Name | 3-sulfanylpropane-1,2-diol |
| Molecular Weight (g/mol) | 108.16 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction