missing translation for 'onlineSavingsMsg'
Learn More

[1-(Tetrahydropyran-2-yl)-1H-pyrazol-5-yl]boronic Acid Pinacol Ester, TRC

Product Code. 30456437
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456437 10 mg 10mg
30451586 50 mg -
30449789 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456437 Supplier Toronto Research Chemicals Supplier No. TRCT88955810MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 903550-26-5
Molecular Formula C14H23BN2O3
Molecular Weight (g/mol) 278.15
Synonym 1-(Tetrahydro-2H-pyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-(Oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, 1-(Oxan-2-yl)-5-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic Acid Pinacol Ester, 1-(Tetrahydropyran-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 1-Tetrahydropyran-2-yl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
IUPAC Name 1-(oxan-2-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole
SMILES CC1(C)OB(OC1(C)C)c2ccnn2C3CCCCO3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H23BN2O3/c1-13(2)14(3,4)20-15(19-13)11-8-9-16-17(11)12-7-5-6-10-18-12/h8-9,12H,5-7,10H2,1-4H3
Formula Weight 278.1802
Chemical Name or Material [1-(Tetrahydropyran-2-yl)-1H-pyrazol-5-yl]boronic Acid Pinacol Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.