missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecyne, tech. 90%

Product Code. 11397745
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11397745 5 g 5g
11307755 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11397745 Supplier Thermo Scientific Alfa Aesar Supplier No. L02573.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 765-10-6 | C14H26 | 194.362 g/mol

1-Tetradecyne, is used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecyne, is used in addition reactions which are typical in alkyne reactions such as halogenation, hydrogenation, hydrohalogenation, hydration, oxidative cleavage, nitrile formation and acidity of terminal alkynes. Polymerisation and substitution reactions are also useful in chemical synthesis.

Solubility
Not miscible with water. soluble in common organic solvents of low polarity

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-10-6
Molecular Formula C14H26
Molecular Weight (g/mol) 194.362
MDL Number MFCD00015076
InChI Key DZEFNRWGWQDGTR-UHFFFAOYSA-N
Synonym 1-tetradecyne, tetradecyne, tetradec-1-in, acmc-1bfxn
PubChem CID 69823
IUPAC Name tetradec-1-yne
SMILES CCCCCCCCCCCCC#C

Specifications

Density 0.79
Boiling Point 132°C to 134°C (14 mmHg)
Refractive Index 1.441
Quantity 5 g
Beilstein 1746546
Solubility Information Not miscible with water. soluble in common organic solvents of low polarity
Formula Weight 194.36
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Tetradecyne

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.