missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecylphosphonic acid, 98%, Thermo Scientific Chemicals

Product Code. 11418127
Click to view available options
Quantity:
1 g
5 g
25 g

1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecylphosphonic acid is used to functionalize the surface of BaTiO3 nanoparticles.

Solubility
Soluble in ethanol, dimethyl sulfoxide and dimethyl formamide. Partially soluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible withstrong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4671-75-4
Molecular Formula C14H31O3P
Molecular Weight (g/mol) 278.373
MDL Number MFCD00015834
InChI Key BVQJQTMSTANITJ-UHFFFAOYSA-N
Synonym phosphonic acid, tetradecyl, 1-tetradecanephosphonic acid, tetradecanephosphonic acid, tetradecyl phosphonic acid, n-tetradecylphosphonic acid, tetradecyl-phosphonic acid, 1-tetradecylphosphonic acid, phosphonic acid, p-tetradecyl, p-tetradecyl-phosphonic acid, tetradecylphosphonate
PubChem CID 78401
IUPAC Name tetradecylphosphonic acid
SMILES CCCCCCCCCCCCCCP(=O)(O)O

Specifications

Quantity 5 g
Solubility Information Soluble in ethanol,dimethyl sulfoxide and dimethyl formamide. Partially soluble in water.
Formula Weight 278.37
Percent Purity 98%
Chemical Name or Material 1-Tetradecylphosphonic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'