missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecylamine, 98%

Product Code. 10215000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
50g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10215000 10 g Glass bottle 10g
10772695 5 g Glass bottle 5g
10045723 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10215000 Supplier Thermo Scientific Acros Supplier No. 138100100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2016-42-4 | C14H31N | 213.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2016-42-4
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.41
InChI Key PLZVEHJLHYMBBY-UHFFFAOYSA-N
Synonym tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d
PubChem CID 16217
IUPAC Name tetradecan-1-amine
SMILES CCCCCCCCCCCCCCN

Specifications

Melting Point 37°C to 42°C
Color Pink to White
Density 0.9930g/mL
Boiling Point 291°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)13NH2
Refractive Index 1.5045 to 1.5065
Quantity 10 g
Beilstein 04, 201
Specific Gravity 0.993
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 213.41
Percent Purity 98%
Physical Form Crystalline Solid or Crystals
Chemical Name or Material 1-Tetradecylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Tetradecylamine
Signal Word
  • Danger
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.