missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylthiuram Disulfide, 97%

Product Code. 10437150 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
2.5kg
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10437150 500 g Plastic bottle 500g
10776151 25 g Plastic bottle 25g
10494545 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10437150 Supplier Thermo Scientific Acros Supplier No. 138125000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 97-77-8 | C10H20N2S4 | 296.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-77-8
Molecular Formula C10H20N2S4
Molecular Weight (g/mol) 296.51
MDL Number MFCD00008157
InChI Key AUZONCFQVSMFAP-UHFFFAOYSA-N
Synonym disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd
PubChem CID 3117
ChEBI CHEBI:4659
IUPAC Name diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 70.0°C to 74.0°C
Color Yellow
Boiling Point 117°C (23.0 hPa)
Loss on Drying 0.3% max. (60°C)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula [(C2H5)2NCS2-]2
Quantity 500 g
Beilstein 04, 122
Fieser 06,569
Merck Index 15, 3407
Solubility Information Solubility in water: 0.02g/100mL. Other solubilities: 3.82g/100mL alcohol, 7.14g/100mL ether, soluble in acetone, benzene, carbon disulfice and, chloroform
Formula Weight 296.51
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Granules
Chemical Name or Material Tetraethylthiuram disulfide, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetraethylthiuram disulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.