missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Tetradecene, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | 1-Tetradecene |
| CAS | 1120-36-1 |
| Analyte or Component Names | 1-Tetradecene |
| Shipping Condition | +20°C |
| Molecular Formula | C14 H28 |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/C14H28/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3H,1,4-14H2,2H3 |
| SMILES | CCCCCCCCCCCCC=C |
| IUPAC Name | tetradec-1-ene |
| Molecular Weight (g/mol) | 196.37 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction