missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecene, 94%

Product Code. 11441728
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441728 2500 mL 2500mL
11404150 100 mL 100mL
11431728 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441728 Supplier Thermo Scientific Alfa Aesar Supplier No. A13974.0F
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1120-36-1 | C14H28 | 196.378 g/mol

1-Tetradecene used in the production of linear plasticizers, oxo-alcohols, motor fuels, lubricants, automotive additives, biodegradable surfactants, paper size, and in a wide range of specialty applications in the production of mercaptans, flavors and fragrances, alkyl metals, halides, alkyl silanes.. It is also used as a co monomers in polymer such as low density polyethylenes for the properties of lighter, thinner, better flexibility and more tearing resistance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Tetradecene used in the production of linear plasticizers, oxo-alcohols, motor fuels, lubricants, automotive additives, biodegradable surfactants, paper size, and in a wide range of specialty applications in the production of mercaptans, flavors and fragrances, alkyl metals, halides, alkyl silanes.. It is also used as a co monomers in polymer such as low density polyethylenes for the properties of lighter, thinner, better flexibility and more tearing resistance.

Solubility
Soluble in hydrocarbon solvents; insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-36-1
Molecular Formula C14H28
Molecular Weight (g/mol) 196.378
MDL Number MFCD00008981
InChI Key HFDVRLIODXPAHB-UHFFFAOYSA-N
Synonym 1-tetradecene, n-tetradec-1-ene, tetradecene, 1-tetradecylene, alpha-tetradecene, unii-fw23481s7s, ccris 3785, alkenes, c10-16 .alpha., a-tetradecene, tetradecene-1
PubChem CID 14260
ChEBI CHEBI:77505
IUPAC Name tetradec-1-ene
SMILES CCCCCCCCCCCCC=C

Specifications

Melting Point -12°C
Density 0.775
Boiling Point 250°C to 252°C
Flash Point 115°C (239°F)
Assay Percent Range 94%
Odor Mild
Linear Formula CH3(CH2)11CH=CH2
Refractive Index 1.436
Quantity 2500 mL
Beilstein 1701156
Solubility Information Soluble in hydrocarbon solvents; insoluble in water.
Formula Weight 196.38
Percent Purity 94%
Chemical Name or Material 1-Tetradecene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.