missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecanol, TRC

Product Code. 17951385
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17951385 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17951385 Supplier Toronto Research Chemicals Supplier No. TRCT29248025G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 112-72-1
Molecular Formula C14 H30 O
Quantity 25 g
Synonym 1-Tetradecanol, Tetradecanol (7CI), 1-Hydroxytetradecane, 1-Tetradecyl alcohol, Adol 18, Alfol 14, Conol 1495, Hainol 14SS, Hinol 14SS, Kalcohl 40, Kalcohl 4098, Kalcol 4098, Kolliwax MA, Lanette 14, Lanette K, Lanette Wax KS, Lorol C 14, Loxanol V, Myristic alcohol, Myristyl alcohol, NSC 8549, Nacol 14-95, Nacol 14-98, Nafol 14, Speziol C 14 Pharma, Tetradecyl alcohol, n-Tetradecan-1-ol, n-Tetradecanol, n-Tetradecyl alcohol
InChI Formula InChI=1S/C14H30O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h15H,2-14H2,1H3
SMILES CCCCCCCCCCCCCCO
IUPAC Name tetradecan-1-ol
Molecular Weight (g/mol) 214.39
Formula Weight 214.2297
Chemical Name or Material 1-n-Tetradecanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.