missing translation for 'onlineSavingsMsg'
Learn More

1-Tetradecanethiol, 94%

Product Code. 11387406
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11387406 5 g 5g
11397406 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11387406 Supplier Thermo Scientific Alfa Aesar Supplier No. L09401.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2079-95-0 | C14H30S | 230.454 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2079-95-0
Molecular Formula C14H30S
Molecular Weight (g/mol) 230.454
MDL Number MFCD00022099
InChI Key GEKDEMKPCKTKEC-UHFFFAOYSA-N
Synonym 1-tetradecanethiol, myristyl mercaptan, tetradecanethiol, n-tetradecyl mercaptan, tetradecyl mercaptan, 1-mercaptotetradecane, unii-apy3p7s1xm, apy3p7s1xm, tetradecanethiol-1, acmc-1cq2y
PubChem CID 64677
IUPAC Name tetradecane-1-thiol
SMILES CCCCCCCCCCCCCCS

Specifications

Melting Point 6°C to 7°C
Density 0.85
Boiling Point 176°C to 180°C (22 mmHg)
Flash Point >110°C (230°F)
Odor Unpleasant
Linear Formula CH3(CH2)13SH
Refractive Index 1.461
Quantity 5 g
Beilstein 1742653
Sensitivity Air Sensitive
Formula Weight 230.46
Percent Purity 94%
Chemical Name or Material 1-Tetradecanethiol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.