missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butylpiperazine, 97%, Thermo Scientific Chemicals

Product Code. 15432779
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38216-72-7
Molecular Formula C8H18N2
Molecular Weight (g/mol) 142.246
MDL Number MFCD00049359
InChI Key PVMNSAIKFPWDQG-UHFFFAOYSA-N
Synonym n-tert-butylpiperazine, 1-tert-butyl piperazine, n-t-butylpiperazine, 1-t-butyl piperazine, 1-tertbutyl piperazine, 1-tert-butyl-piperazine, piperazine, 1-1,1-dimethylethyl, n-tert-butyl piperazine, tert-butylpiperazine, tert-butyl piperazine
PubChem CID 3530572
IUPAC Name 1-tert-butylpiperazine
SMILES CC(C)(C)N1CCNCC1

Specifications

Melting Point 32°C to 35°C
Color Colorless to Yellow
Boiling Point 85°C to 87°C (22 mmHg)
Quantity 1 g
UN Number UN3259
Sensitivity Air, light, and moisture sensitive
Formula Weight 142.24
Percent Purity 97%
Physical Form Low Melting Solid
Chemical Name or Material 1-tert-Butylpiperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'