missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-butyl)-5-methyl-1H-pyrazole-3-carbohydrazide, Thermo Scientific™

Product Code. 10168054
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10168054 5 g 5g
10631123 250 mg 250mg
10724023 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10168054 Supplier Thermo Scientific Maybridge Supplier No. SPB08431DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306937-23-5
Molecular Formula C9H16N4O
Molecular Weight (g/mol) 196.254
InChI Key TWKKUKBETOOWRR-UHFFFAOYSA-N
Synonym 1-tert-butyl-5-methyl-1h-pyrazole-3-carbohydrazide, maybridge1_008666, 1-tert-butyl-5-methyl-pyrazole-3-carbohydrazide, 1-tert-butyl-5-methylpyrazole-3-carbohydrazide, 1h-pyrazole-3-carboxylicacid, 1-1,1-dimethylethyl-5-methyl-, hydrazide
PubChem CID 2735685
IUPAC Name 1-tert-butyl-5-methylpyrazole-3-carbohydrazide
SMILES CC1=CC(=NN1C(C)(C)C)C(=O)NN

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 196.25
Percent Purity 97%
Chemical Name or Material 1-(tert-Butyl)-5-methyl-1H-pyrazole-3-carbohydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.