missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl 4-(9-H-fluoren-9-ylmethyl) hydrogen (2S)-piperazine-1,2,4-tricarboxyl, 97%, Maybridge™

Brand:  Maybridge MO08424DA

Code : 31

Additional Details : CAS Number : 1034574-30-5 Weight : 0.05100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Packaging Amber Glass Bottle
Quantity 1g

 View more versions of this product

Product Code. 10031624

  • £172.00 / 1g

Estimated Shipment: 20-08-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1034574-30-5
452.507
ZVHNNCSUTNWKFC-NRFANRHFSA-N
7128293
CC(C)(C)OC(=O)N1CCN(CC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
C25H28N2O6
MFCD02179101
s-1-n-boc-4-n-fmoc-piperazine-2-carboxylic acid, s-1-n-boc-4-n-fmoc-piperazine2-carboxylic acid, s-4-9h-fluoren-9-yl methoxy carbonyl-1-tert-butoxycarbonyl piperazine-2-carboxylic acid, s-1-n-boc-4-n-fmoc-2-piperazine carboxylic acid, 1-boc-4-fmoc-piperazine-2-s-carboxylic acid, 2s-1-tert-butoxycarbonyl-4-9h-fluoren-9-ylmethoxy carbonyl piperazine-2-carboxylic acid, s-1-n-boc-4-n-fmoc-piperazine 2-carboxylic acid, s-piperazine-1,2,4-tricarboxylic acid 1-tert-butyl ester 4-9h-fluoren-9-ylmethyl ester, 1-tert-butyl 4-9-h-fluoren-9-ylmethyl hydrogen 2s-piperazine-1,2,4-tricarboxylate, bestipharma 114-505
(2S)-4-(9H-fluoren-9-ylmethoxycarbonyl)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperazine-2-carboxylic acid

Specifications

1-tert-Butyl 4-(9-H-fluoren-9-ylmethyl) hydrogen (2S)-piperazine-1, 2, 4-tricarboxyl
1g
97.0
C25H28N2O6
s-1-n-boc-4-n-fmoc-piperazine-2-carboxylic acid, s-1-n-boc-4-n-fmoc-piperazine2-carboxylic acid, s-4-9h-fluoren-9-yl methoxy carbonyl-1-tert-butoxycarbonyl piperazine-2-carboxylic acid, s-1-n-boc-4-n-fmoc-2-piperazine carboxylic acid, 1-boc-4-fmoc-piperazine-2-s-carboxylic acid, 2s-1-tert-butoxycarbonyl-4-9h-fluoren-9-ylmethoxy carbonyl piperazine-2-carboxylic acid, s-1-n-boc-4-n-fmoc-piperazine 2-carboxylic acid, s-piperazine-1,2,4-tricarboxylic acid 1-tert-butyl ester 4-9h-fluoren-9-ylmethyl ester, 1-tert-butyl 4-9-h-fluoren-9-ylmethyl hydrogen 2s-piperazine-1,2,4-tricarboxylate, bestipharma 114-505
CC(C)(C)OC(=O)N1CCN(CC1C(=O)O)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
452.507
452.5
Amber Glass Bottle
1034574-30-5
97%
MFCD02179101
ZVHNNCSUTNWKFC-NRFANRHFSA-N
(2S)-4-(9H-fluoren-9-ylmethoxycarbonyl)-1-[(2-methylpropan-2-yl)oxycarbonyl]piperazine-2-carboxylic acid
7128293
97%
Videos
SDS Documents
Certificates