missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid, ≥95%, Maybridge™

Brand:  Maybridge SPB03128DA

Code : 31

Additional Details : CAS Number : 175277-09-5 Weight : 0.05100kg

Quantity 1g

 View more versions of this product

Product Code. 10008171

  • £40.18 / 1g

Estimated Shipment
01-06-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

175277-09-5
182.223
JZPMLZWJUMATOQ-UHFFFAOYSA-N
2744573
CC1=NN(C(=C1)C(=O)O)C(C)(C)C
C9H14N2O2
MFCD00084917
1-tert-butyl-3-methyl-1h-pyrazole-5-carboxylic acid, 2-tert-butyl-5-methyl-pyrazole-3-carboxylic acid, 1-tert-butyl-3-methylpyrazole-5-carboxylic acid, maybridge1_008627, buttpark 9218-70, 2-tert-butyl-5-methyl-3-pyrazolecarboxylic acid, 1-tert-butyl-5-carboxy-3-methyl-1h-pyrazole, 1-tert-butyl-3-methyl-1h-pyrazole-5-carboxylic aci
2-tert-butyl-5-methylpyrazole-3-carboxylic acid

Specifications

1-(tert-Butyl)-3-methyl-1H-pyrazole-5-carboxylic acid
1g
97.0
C9H14N2O2
1-tert-butyl-3-methyl-1h-pyrazole-5-carboxylic acid, 2-tert-butyl-5-methyl-pyrazole-3-carboxylic acid, 1-tert-butyl-3-methylpyrazole-5-carboxylic acid, maybridge1_008627, buttpark 9218-70, 2-tert-butyl-5-methyl-3-pyrazolecarboxylic acid, 1-tert-butyl-5-carboxy-3-methyl-1h-pyrazole, 1-tert-butyl-3-methyl-1h-pyrazole-5-carboxylic aci
CC1=NN(C(=C1)C(=O)O)C(C)(C)C
182.223
182.22
Amber glass bottle
175277-09-5
97%
MFCD00084917
JZPMLZWJUMATOQ-UHFFFAOYSA-N
2-tert-butyl-5-methylpyrazole-3-carboxylic acid
2744573
97%
Videos
SDS Documents