missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1-tert-Butyl-3,5-dimethyl-1H-pyrazole-4-carboxaldehyde, 95%

Product Code. 11395539
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11395539 1 g 1g
11305549 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395539 Supplier Alfa Aesar™ Supplier No. H50480.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 647824-51-9
Molecular Formula C10H16N2O
Molecular Weight (g/mol) 180.251
MDL Number MFCD00120293
InChI Key WQHXOHHEPNBXMF-UHFFFAOYSA-N
Synonym 1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carbaldehyde, 1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carboxaldehyde, buttpark 99\12-15, 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde, acmc-20ap9c
PubChem CID 2776936
IUPAC Name 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde
SMILES CC1=C(C(=NN1C(C)(C)C)C)C=O

Specifications

Quantity 1 g
Formula Weight 180.25
Percent Purity 95%
Chemical Name or Material 1-tert-Butyl-3,5-dimethyl-1H-pyrazole-4-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.