missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-butyl) 2-methyl 1H-indole-1,2-dicarboxylate, 97%, Thermo Scientific™

Product Code. 10577683
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10577683 1 g 1g
10381343 250 mg 250mg
10717875 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10577683 Supplier Thermo Scientific Maybridge Supplier No. CC27822DA
 Restricted Item
Estimated Shipment: 04-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 163229-48-9
Molecular Formula C15H17NO4
Molecular Weight (g/mol) 275.304
InChI Key UJVYYFYNECMKRA-UHFFFAOYSA-N
Synonym 1-tert-butyl 2-methyl 1h-indole-1,2-dicarboxylate, 1-tert-butyl 2-methyl indole-1,2-dicarboxylate, 1-tert-butyl-2-methyl-1h-indole-1,2-dicarboxylate, 1h-indole-1,2-dicarboxylic acid 1-tert-butyl 2-methyl ester, 1h-indole-1,2-dicarboxylicacid, 1-1,1-dimethylethyl 2-methyl ester
PubChem CID 2795473
IUPAC Name 1-O-tert-butyl 2-O-methyl indole-1,2-dicarboxylate
SMILES CC(C)(C)OC(=O)N1C2=CC=CC=C2C=C1C(=O)OC

Specifications

Assay Percent Range 97%
Quantity 1 g
Formula Weight 275.3
Percent Purity 97%
Chemical Name or Material 1-(tert-Butyl) 2-methyl 1H-indole-1, 2-dicarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.