missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(tert-Butoxycarbonyl)-4-oxopyrrolidine-2-carboxylic acid 97+%, Thermo Scientific™
£23.00 - £122.00
Chemical Identifiers
CAS | 84348-37-8 |
---|---|
Molecular Formula | C10H15NO5 |
Molecular Weight (g/mol) | 229.23 |
MDL Number | MFCD01860669 |
InChI Key | CKYGSXRXTIKGAJ-UHFFFAOYNA-N |
Synonym | n-boc-4-oxo-l-proline, s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, n-t-boc-4-oxo-l-proline, boc-4-oxo-pro-oh, n-tert-boc-4-oxo-l-proline, 2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-tert-butoxycarbonyl-4-oxoproline, 2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester |
PubChem CID | 11593804 |
IUPAC Name | (2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopyrrolidine-2-carboxylic acid |
SMILES | CC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10245604
|
Thermo Scientific Maybridge
MO07783CB |
250 mg | Amber Glass Bottle |
£23.00
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10316904
|
Thermo Scientific Maybridge
MO07783DA |
1 g | Amber Glass Bottle |
£43.75
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10421504
|
Thermo Scientific Maybridge
MO07783DE |
5 g | Amber Glass Bottle |
£122.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
84348-37-8 | |
229.23 | |
CKYGSXRXTIKGAJ-UHFFFAOYNA-N | |
11593804 | |
CC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O |
C10H15NO5 | |
MFCD01860669 | |
n-boc-4-oxo-l-proline, s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, n-t-boc-4-oxo-l-proline, boc-4-oxo-pro-oh, n-tert-boc-4-oxo-l-proline, 2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-tert-butoxycarbonyl-4-oxoproline, 2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester | |
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopyrrolidine-2-carboxylic acid |
Specifications
84348-37-8 | |
C10H15NO5 | |
n-boc-4-oxo-l-proline, s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, n-t-boc-4-oxo-l-proline, boc-4-oxo-pro-oh, n-tert-boc-4-oxo-l-proline, 2s-1-tert-butoxycarbonyl-4-oxopyrrolidine-2-carboxylic acid, 1-tert-butoxycarbonyl-4-oxoproline, 2s-1-tert-butoxycarbonyl-4-oxo-pyrrolidine-2-carboxylic acid, 2s-1-tert-butoxy carbonyl-4-oxopyrrolidine-2-carboxylic acid, s-4-oxo-pyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester | |
CC(C)(C)OC(=O)N1CC(=O)CC1C(O)=O | |
229.23 | |
229.23 | |
(2S)-1-tert-butyl hydrogen 4-oxopyrrolidine-1, 2-dicarboxylate |
97% | |
MFCD01860669 | |
CKYGSXRXTIKGAJ-UHFFFAOYNA-N | |
(2S)-1-[(2-methylpropan-2-yl)oxycarbonyl]-4-oxopyrrolidine-2-carboxylic acid | |
11593804 | |
97% |
Safety and Handling
GHS P Statement
Harmful if swallowed.
Causes serious eye irritation.
GHS P Statement
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.