missing translation for 'onlineSavingsMsg'
Learn More

1-(tert-Butoxycarbonyl)-2-pyrrolidinone, 97%

Product Code. 17251222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17251222 5 g 5g
17151852 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17251222 Supplier Thermo Scientific Acros Supplier No. 466330050
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 85909-08-6 | C9H15NO3 | 185.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85909-08-6
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.22
InChI Key GJJYYMXBCYYXPQ-UHFFFAOYSA-N
IUPAC Name tert-butyl 2-oxopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1=O

Specifications

Density 1.0860 g/mL
Assay Percent Range 97%
Quantity 5 g
Specific Gravity 1.086
Formula Weight 185.22
Chemical Name or Material 1-(tert-Butoxycarbonyl)-2-pyrrolidinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.