missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ 1-step Fix/Lyse Solution (10X)

Enables lysis of RBCs after staining peripheral blood cells with fluorochrome-conjugated antibodies. Formulated to lyse non-nucleated erythrocytes while maintaining fixed and labeled leukocyte population.

£75.00 - £238.00

Alert:

Contains 10% formaldehyde and 30% diethylene glycol


Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15596286
View Documents
Invitrogen™
00-5333-54
50mL £75.00
50mL View alternative products
Save Up to 
»
In stock, ready to ship: 3
Add to basket
 
15506296
View Documents
Invitrogen™
00-5333-57
200mL £238.00
200mL View alternative products
Save Up to 
»
In stock, ready to ship: 3
Add to basket
 
Description

Description

  • Isolation of mononuclear cells by gradient centrifugation (e.g. Ficoll-Paque density gradient separation) is not required when using this solution
  • Compatible with antibodies conjugated to organic dyes and tandem dyes

For Research Use Only