missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uvex 1 Sport White NC Shoe O1 FO SRC, Width 11
Shop All Uvex Products
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
49
50
51
52
Unit Size:
1 pair
Description
- Modern, ultra-lightweight and flexible O1 occupational shoe without protective toe cap
- Washable at up to 30°C
- Made from synthetic materials, so suitable for people allergic to chrome
- Elastic laces with quick release for individual adjustment
- All sole materials are free of silicone, plasticizers and other substances that interfere with wetting agents
- New, innovative uvex i-PUREnrj sole technology provides outstanding shock-absorption properties at the forefoot and heel and excellent rebound energy over the entire midsole
- Meets the ESD specification with volume resistance less than 35 Megohms
- Ergonomically designed outsole made from dual density polyurethane with excellent slip resistance
- Outstanding wearer comfort thanks to a newly developed last and climate-optimized, breathable materials
- Virtually seam-free construction with high-tech materials to eliminate pressure points
- Removable anti-static comfortable insole with a moisture transport system and additional shock absorption in the forefoot and heel
- Soft padding on collar and tongue
- Made partly from recycled materials in order to conserve resources
- Free from any of the harmful substances listed on our banned hazardous substances list
Specifications
Specifications
| Color | White |
| Material | Textile |
| Size | 38 |
| Toe Material | Plain Toe |
| Certifications/Compliance (Europe) | EN ISO 20347:2012 O1 FO SRC EN ISO 20347:2022 O1 FO SC SR |
| Gender | Unisex |
| Fit | Standard Width |
| Latex Free | Yes |
| Ankle Style | Trainers |
| Waterproof | No |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction