Learn More
Agilent Technologies 1 SET Rack overlay for 40-position sample rack for
Brand:  Agilent Technologies 6610026000
Encompass_Preferred
1 SET Rack overlay for 40-position sample rack for SPS 3 autosampler allow s 40 x 18 mm od tube
Product Code. 30187452
  • £106.65 / 1 set
Only null left
Estimated Shipment: 07-01-2026