missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human ErbB3/Her3 Quantikine ELISA Kit
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
Specifications
Specifications
| Assay Sensitivity | 15.2 pg/mL |
| Specificity | Natural and Recombinant Human ErbB3 |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 2065 |
| Measuring Range | 62.50 to 4000 pg/mL |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) |
| Target | ErbB3/Her3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction