missing translation for 'onlineSavingsMsg'
Learn More

1-S-Octyl-β-D-thioglucopyranoside, 98%, Thermo Scientific Chemicals

Product Code. p-176722 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
500mg
This item is not returnable. View return policy

Product Code. 10731361

Brand: Thermo Scientific Acros 315005000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85618-21-9
Molecular Formula C14H28O5S
Molecular Weight (g/mol) 308.44
MDL Number MFCD00012189
InChI Key CGVLVOOFCGWBCS-DGTMBMJNSA-N
Synonym octyl thioglucopyranoside, octyl-i 2-d-1-thioglucopyranoside
PubChem CID 53720324
IUPAC Name (2R,3S,4S,5S,6S)-2-(hydroxymethyl)-6-octylsulfanyloxane-3,4,5-triol
SMILES CCCCCCCCSC1C(C(C(C(O1)CO)O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 500 mg
Specific Rotation Condition − 46.00 (20.00°C c=1,CH3OH)
Specific Rotation -46
Solubility Information Solubility in water: soluble
Formula Weight 308.44
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 1-S-Octyl-β-D-thioglucopyranoside, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.