missing translation for 'onlineSavingsMsg'
Learn More

(1'RS,2'S)-Nicotine 1,1'-Di-N-Oxide [20% in ethanol], TRC

Product Code. 30479538
Change view
Click to view available options
Quantity:
0.5 mL
5 mL
Unit Size:
0.5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30479538 0.5 mL 0.5mL
30465490 5 mL -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30479538 Supplier Toronto Research Chemicals Supplier No. TRCN4150000.5ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129547-84-8
Molecular Formula C10 H14 N2 O2
Molecular Weight (g/mol) 194.23
Synonym (1Rs,2S)-Nicotine 1,1-Di-N-Oxide
IUPAC Name 3-[(2S)-1-methyl-1-oxidopyrrolidin-1-ium-2-yl]-1-oxidopyridin-1-ium
SMILES C[N+]1([O-])CCC[C@H]1c2ccc[n+]([O-])c2

Specifications

Quantity 0.5 mL
InChI Formula InChI=1S/C10H14N2O2/c1-12(14)7-3-5-10(12)9-4-2-6-11(13)8-9/h2,4,6,8,10H,3,5,7H2,1H3/t10-,12?/m0/s1
Formula Weight 194.1055
Concentration or Composition (by Analyte or Components) 20%
Chemical Name or Material (1'R, 2'S)-Nicotine 1,1'-Di-N-Oxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.