missing translation for 'onlineSavingsMsg'
Learn More

(1’R,2S)-2-(1’,2’-Dihydroxyethyl)-6-fluorochromane, TRC

Product Code. 30382040
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382040 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30382040 Supplier Toronto Research Chemicals Supplier No. TRCD449038100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (1'R,2S)-2-(1',2'-Dihydroxyethyl)-6-fluorochromane
CAS 303176-39-8
Molecular Formula C11 H13 F O3
Quantity 100 mg
Synonym (1R)-1-[(2S)-6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-yl]-1,2-ethanediol
InChI Formula InChI=1S/C11H13FO3/c12-8-2-4-10-7(5-8)1-3-11(15-10)9(14)6-13/h2,4-5,9,11,13-14H,1,3,6H2/t9-,11+/m1/s1
SMILES OC[C@@H](O)[C@@H]1CCc2cc(F)ccc2O1
IUPAC Name (1R)-1-[(2S)-6-fluoro-3,4-dihydro-2H-chromen-2-yl]ethane-1,2-diol
Molecular Weight (g/mol) 212.218
Formula Weight 212.085
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.