missing translation for 'onlineSavingsMsg'
Learn More

1-Pyrrolidinecarbonyl chloride, 97%

Product Code. 11343027
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11343027 1 g 1g
11353027 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11343027 Supplier Thermo Scientific Alfa Aesar Supplier No. L13396.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1192-63-8 | C5H8ClNO | 133.575 g/mol

1-Pyrrolidinecarbonyl chloride, is used in the synthesis of (S)-4-(2-(((benzyloxy)carbonyl)amino)-3-methoxy-3-oxopropyl)phenyl pyrrolidine-1-carboxylate

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Pyrrolidinecarbonyl chloride, is used in the synthesis of (S)-4-(2-(((benzyloxy)carbonyl)amino)-3-methoxy-3-oxopropyl)phenyl pyrrolidine-1-carboxylate

Solubility
It reacts with water.

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents, bases. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-63-8
Molecular Formula C5H8ClNO
Molecular Weight (g/mol) 133.575
MDL Number MFCD00051321
InChI Key XACWJIQLDLUFSR-UHFFFAOYSA-N
Synonym 1-pyrrolidinecarbonyl chloride, pyrrolidinecarbonyl chloride, 1-pyrrolidinecarbonylchloride, pyrrolidine-1-carbonylchloride, pyrrolidinocarbonyl chloride, 1pyrrolidinecarbonyl chloride, pyrrolidine carbonyl chloride, acmc-1bo54, 1-pyrrolidine carbonylchloride, 1-pyrolidine carbonyl chloride
PubChem CID 70937
IUPAC Name pyrrolidine-1-carbonyl chloride
SMILES C1CCN(C1)C(=O)Cl

Specifications

Boiling Point 75°C (1mmHg)
Refractive Index 1.494
UN Number UN3265
Beilstein 110281
Solubility Information It reacts with water.
Density 1.209
Quantity 1 g
Flash Point 97°C (206°F)
Formula Weight 133.58
Sensitivity Moisture sensitive
Percent Purity 97%
Chemical Name or Material 1-Pyrrolidinecarbonyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 1-Pyrrolidinecarbonyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: 1-Pyrrolidinecarbonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.